Merck
Mitotic DNA Synthesis Is Differentially Regulated between Cancer and Noncancerous Cells.
Molecular cancer research : MCR (2019-05-23)
Cari L Graber-Feesl, Kayla D Pederson, Katherine J Aney, Naoko Shima
ZUSAMMENFASSUNG
Mitotic DNA synthesis is a recently discovered mechanism that resolves late replication intermediates, thereby supporting cell proliferation under replication stress. This unusual form of DNA synthesis occurs in the absence of RAD51 or BRCA2, which led to the identification of RAD52 as a key player in this process. Notably, mitotic DNA synthesis is predominantly observed at chromosome loci that colocalize with FANCD2 foci. However, the role of this protein in mitotic DNA synthesis remains largely unknown. In this study, we investigated the role of FANCD2 and its interplay with RAD52 in mitotic DNA synthesis using aphidicolin as a universal inducer of this process. After examining eight human cell lines, we provide evidence for FANCD2 rather than RAD52 as a fundamental supporter of mitotic DNA synthesis. In cancer cell lines, FANCD2 exerts this role independently of RAD52. Surprisingly, RAD52 is dispensable for mitotic DNA synthesis in noncancerous cell lines, but these cells strongly depend on FANCD2 for this process. Therefore, RAD52 functions selectively in cancer cells as a secondary regulator in addition to FANCD2 to facilitate mitotic DNA synthesis. As an alternative to aphidicolin, we found partial inhibition of origin licensing as an effective way to induce mitotic DNA synthesis preferentially in cancer cells. Importantly, cancer cells still perform mitotic DNA synthesis by dual regulation of FANCD2 and RAD52 under such conditions. IMPLICATIONS: These key differences in mitotic DNA synthesis between cancer and noncancerous cells advance our understanding of this mechanism and can be exploited for cancer therapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium – hoher Glucosegehalt, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RIPA-Puffer
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium/Nährstoffmischung F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
McCoy 5A-Medium, Modified, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Rabbit anti-ORC1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.