Merck
  • In vitro evaluation of gel-encapsulated adipose derived stem cells: Biochemical cues for in vivo peripheral nerve repair.
In vitro evaluation of gel-encapsulated adipose derived stem cells: Biochemical cues for in vivo peripheral nerve repair.
Journal of tissue engineering and regenerative medicine (2017-05-31)
A C de Luca, C M Fonta, W Raffoul, P G di Summa, S P Lacour
ZUSAMMENFASSUNG
Adipose-derived stem cells (ASC) are becoming one of the most exploited cells in peripheral nerve repair. They are fast-growing and able to protect neurons from apoptosis; they can reduce post-injury latency and the risk of muscle atrophy. This study evaluates laminin-loaded fibrin gel as an ASC-carrying scaffold for nerve repair. In vitro, ASC retained their proliferative activity but showed significant increase in proliferation rate when encapsulated in gels with low laminin concentrations (i.e., 1 μg/mL). We observed a linear decrease of ASC proliferation rate with increasing laminin concentration from 1 to 100 μg/mL. We next examined the effect of the ASC-carrying fibrin gels on in vitro dorsal root ganglia (DRG) neurite extension, then in vivo sciatic nerve regeneration in adult rats. The ASC-carrying gel was embedded in 15-mm-long, 1.5-mm-diameter polydimethylsiloxane regenerative conduits for in vivo evaluation. At 8-week post implantation, robust regeneration was observed across the long gap. Taken together, these results suggest ASC-carrying gels are a potential path to improve the efficacy of nerve regeneration through artificial guidance conduits and electrode nerve interfaces.