Merck
  • EPHA4 regulates vascular smooth muscle cell contractility and is a sex-specific hypertension risk gene in individuals with type 2 diabetes.
EPHA4 regulates vascular smooth muscle cell contractility and is a sex-specific hypertension risk gene in individuals with type 2 diabetes.
Journal of hypertension (2019-03-01)
Zeqin Zhang, Johanne Tremblay, John Raelson, Tamar Sofer, Lizhong Du, Qiang Fang, Maria Argos, Francois-Christophe Marois-Blanchet, Yu Wang, Lingling Yan, John Chalmers, Mark Woodward, Stephen Harrap, Pavel Hamet, Hongyu Luo, Jiangping Wu
ZUSAMMENFASSUNG
We investigated the association of genetic variants of EPHA4, a receptor tyrosine kinase, with hypertension, and its role in vascular smooth muscle cell (VSMC) contractility. Data from two human genetic studies, ADVANCE and HCHS/SOL, were analyzed for association of EPHA4 single nucleotide variants (SNVs) with hypertension risks. The effect of EPHA4 signalling on mouse VSMC contractility was assessed. We identified a SNV (rs75843691 hg19 chr2:g.222395371 C>G), located in the third intron of EPHA4 gene, being significantly associated with hypertension in human female patients (P value = 8.3 × 10, below the Bonferroni-corrected critical P value) but not male patients with type 2 diabetes from the ADVANCE clinical trial. We found that EPHA4 was expressed in VSMCs and its stimulation by anti-EPHA4 antibody led to reduced VSMC contractility. Estrogen enhanced the contractility-lowering effect of EPHA4 stimulation. Conversely, siRNA knockdown of Epha4 expression in VSMCs resulted in increased contractility of VSMCs from female mice but not from male mice. EPHA4 appears to be a sex-specific hypertension risk gene in type 2 diabetic patients. Forward EPHA4 signalling reduces VSMC contractility, and estrogen is a modifier of this effect. The effect of EPHA4 on VSMCs contractility explains the association of EPHA4 gene with hypertension risks in female patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.