Merck
  • Protamine-templated biomimetic hybrid capsules: efficient and stable carrier for enzyme encapsulation
Protamine-templated biomimetic hybrid capsules: efficient and stable carrier for enzyme encapsulation
Chemistry of Materials (2007)
Zhang Y, et al.