Merck
  • Physical and enzymological interaction of Bacillus subtilis proteins required for de novo pyridoxal 5?-phosphate biosynthesis
Physical and enzymological interaction of Bacillus subtilis proteins required for de novo pyridoxal 5?-phosphate biosynthesis
Journal of Bacteriology (2004)
Belitsky BR