Merck
MicroRNA-628-5p inhibits cell proliferation in glioma by targeting DDX59.
Journal of cellular biochemistry (2019-05-22)
Peng Xie, Yan Wang, Yuanmei Liao, Qiu Han, Zhichao Qiu, Yinbing Chen, Xiaohua Zuo
ZUSAMMENFASSUNG
Recent study has reported that microRNA-628-5p (miR-628-5p) is involved in the development of epithelial ovarian cancer; however, the mechanisms of miR-628-5p in glioma remain unclear. In this study, we explored the potential biological roles of miR-628-5p in glioma. First, we found that miR-628-5p was decreased in the tissues and cells (U87 and T98) of glioma. Second, overexpressing miR-628-5p reduced the ability of glioma cells' proliferation and induced glioma cells' cycle arrest in G1. Then, we found that miR-628-5p directly bound to the 3'-untranslated region of DDX59 and decreased the protein level of DDX59. The decrease of DDX59 was found to lead to the decrease of p-AKT. Mechanistic studies revealed that restoring the expression of DDX59 alleviated miR-628-5p-induced inhibition of proliferation of glioma. These findings suggest that the miR-628-5p/DDX59 axis has a key role in the development of glioma, and miR-628-5p might be a new therapeutic target against glioma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human DDX59
Preise und Verfügbarkeit sind derzeit nicht verfügbar.