- Rapid, large-volume, thermally controlled 3D printing using a mobile liquid interface.
Rapid, large-volume, thermally controlled 3D printing using a mobile liquid interface.
Science (New York, N.Y.) (2019-10-19)
David A Walker, James L Hedrick, Chad A Mirkin
PMID31624211
ZUSAMMENFASSUNG
We report a stereolithographic three-dimensional printing approach for polymeric components that uses a mobile liquid interface (a fluorinated oil) to reduce the adhesive forces between the interface and the printed object, thereby allowing for a continuous and rapid print process, regardless of polymeric precursor. The bed area is not size-restricted by thermal limitations because the flowing oil enables direct cooling across the entire print area. Continuous vertical print rates exceeding 430 millimeters per hour with a volumetric throughput of 100 liters per hour have been demonstrated, and proof-of-concept structures made from hard plastics, ceramic precursors, and elastomers have been printed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.