Merck
Sustainable ecofriendly phytoextract mediated one pot green recovery of chitosan.
Scientific reports (2019-09-27)
Judy Gopal, Manikandan Muthu, Thirumalai Dhakshanamurthy, Ki Jun Kim, Nazim Hasan, Seong Jung Kwon, Sechul Chun
ZUSAMMENFASSUNG
Chitin and chitosan are biopolymers that have diverse applications in medicine, agriculture, food, cosmetics, pharmaceuticals, wastewater treatment and textiles. With bio-origins, they easily blend with biological systems and show exemplified compatibility which is mandatory when it comes to biomedical research. Chitin and chitosan are ecofriendly, however the processes that are used to recover them aren't ecofriendly. The focus of this work is to attempt an ecofriendly, sustainable phytomediated one pot recovery of chitosan from commercial chitin and from crab and shrimp shells and squid pen solid wastes. Graviola extracts have been employed, given the fact file that their active ingredients acetogenins actively interact with chitin in insects (resulting in its application as an insecticide). With that as the core idea, the graviola extracts were chosen for orchestrating chitin recovery and a possible chitin to chitosan transformation. The results confirm that graviola extracts did succeed in recovery of chitosan nanofibers from commercial chitin flakes and recovery of chitosan particles directly from solid marine wastes of crab, shrimp and squids. This is a first time report of a phyto-extract mediated novel chitosan synthesis method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chitin aus Garnelenschalen, practical grade, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.