Merck
  • Bioactive milk proteins, peptides and lipids and other functional components derived from milk and bovine colostrum
Bioactive milk proteins, peptides and lipids and other functional components derived from milk and bovine colostrum
Functional Foods (2011)