- In vitro activities of temafloxacin (A-62254) and four other antibiotics against Chlamydia trachomatis.
In vitro activities of temafloxacin (A-62254) and four other antibiotics against Chlamydia trachomatis.
Antimicrobial agents and chemotherapy (1989-01-01)
J Segreti, H A Kessler, K S Kapell, G M Trenholme
PMID2712543
ZUSAMMENFASSUNG
The in vitro activity of temafloxacin (A-62254), a new quinolone antibiotic, against 13 strains of Chlamydia trachomatis was determined and compared with those of doxycycline, norfloxacin, ciprofloxacin, and difloxacin. Temafloxacin and difloxacin were the most active quinolones tested, with bactericidal activity comparable to that of doxycycline.