Merck
  • The heat-shock protein 90 inhibitor 17-allylamino-17-demethoxygeldanamycin suppresses glial inflammatory responses and ameliorates experimental autoimmune encephalomyelitis.
The heat-shock protein 90 inhibitor 17-allylamino-17-demethoxygeldanamycin suppresses glial inflammatory responses and ameliorates experimental autoimmune encephalomyelitis.
Journal of neurochemistry (2006-10-27)
Cinzia Dello Russo, Paul E Polak, Pilar R Mercado, Alessandra Spagnolo, Anthony Sharp, Patricia Murphy, Adeela Kamal, Francis J Burrows, Lawrence C Fritz, Douglas L Feinstein
ZUSAMMENFASSUNG
The heat-shock response (HSR), a highly conserved cellular response, is characterized by rapid expression of heat-shock proteins (HSPs), and inhibition of other synthetic activities. The HSR can attenuate inflammatory responses, via suppression of transcription factor activation. A HSR can be induced pharmacologically by HSP90 inhibitors, through activation of the transcription factor Heat Shock Factor 1 (HSF1). In the present study we characterized the effects of 17-allylamino-17-demethoxygeldanamycin (17-AAG), a less toxic derivative of the naturally occurring HSP90 inhibitor geldanamycin, on glial inflammatory responses and the development of experimental autoimmune encephalomyelitis. In primary enriched glial cultures, 17-AAG dose dependently reduced lipopolysaccharide-dependent expression and activity of inducible nitric oxide synthase, attenuated interleukin (IL)-1beta expression and release, increased inhibitor of kappaB protein levels, and induced HSP70 expression. 17-AAG administration to mice immunized with myelin oligodendrocyte glycoprotein peptide prevented disease onset when given at an early time, and reduced clinical symptoms when given during ongoing disease. T cells from treated mice showed a reduced response to immunogen re-stimulation, and 17-AAG reduced CD3- and CD28-dependent IL-2 production. Together, these data suggest that HSP90 inhibitors could represent a new approach for therapeutic intervention in autoimmune diseases such as multiple sclerosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.