Merck
  • The N termini of focal adhesion kinase family members regulate substrate phosphorylation, localization, and cell morphology.
The N termini of focal adhesion kinase family members regulate substrate phosphorylation, localization, and cell morphology.
The Journal of biological chemistry (2002-09-12)
Jill M Dunty, Michael D Schaller
ZUSAMMENFASSUNG
The focal adhesion kinase (FAK) and cell adhesion kinase beta (CAKbeta, PYK2, CADTK, RAFTK) are highly homologous FAK family members, yet clearly have unique roles in the cell. Comparative analyses of FAK and CAKbeta have revealed intriguing differences in their activities. These differences were investigated further through the characterization of a set of FAK/CAKbeta chimeric kinases. CAKbeta exhibited greater catalytic activity than FAK in vitro, providing a molecular basis for differential substrate phosphorylation by FAK and CAKbeta in vivo. Furthermore, the N terminus may regulate catalytic activity since chimeras containing the FAK N terminus and CAKbeta catalytic domain exhibited a striking high level of catalytic activity and substrate phosphorylation. Unexpectedly, a modulatory role for the N termini in subcellular localization was also revealed. Chimeras containing the FAK N terminus and CAKbeta C terminus localized to focal adhesions, whereas chimeras containing the N and C termini of CAKbeta did not. Finally, prominent changes in cell morphology were induced upon expression of chimeras containing the CAKbeta N terminus, which were not associated with apoptotic cell death, cell cycle progression delay, or changes in Rho activity. These results demonstrate novel regulatory roles for the N terminus of FAK family kinases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-c-Myc-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 9E10, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-avian Src Antibody, clone EC10, clone EC10, Upstate®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.