Merck
  • Allyl-Nickel Catalysis Enables Carbonyl Dehydrogenation and Oxidative Cycloalkenylation of Ketones.
Allyl-Nickel Catalysis Enables Carbonyl Dehydrogenation and Oxidative Cycloalkenylation of Ketones.
Journal of the American Chemical Society (2019-03-13)
David Huang, Suzanne M Szewczyk, Pengpeng Zhang, Timothy R Newhouse
ZUSAMMENFASSUNG
We herein disclose the first report of a first-row transition metal-catalyzed α,β-dehydrogenation of carbonyl compounds using allyl-nickel catalysis. This development overcomes several limitations of previously reported allyl-palladium-catalyzed oxidation, and is further leveraged for the development of an oxidative cycloalkenylation reaction that provides access to bicycloalkenones with fused, bridged, and spirocyclic ring systems using unactivated ketone and alkene precursors.