Merck
  • Abl deregulates Cdk5 kinase activity and subcellular localization in Drosophila neurodegeneration.
Abl deregulates Cdk5 kinase activity and subcellular localization in Drosophila neurodegeneration.
Cell death and differentiation (2006-08-26)
H Lin, T-Y Lin, J-L Juang
ZUSAMMENFASSUNG
Although Abl functions in mature neurons, work to date has not addressed Abl's role on Cdk5 in neurodegeneration. We found that beta-amyloid (Abeta42) initiated Abl kinase activity and that blockade of Abl kinase rescued both Drosophila and mammalian neuronal cells from cell death. We also found activated Abl kinase to be necessary for the binding, activation, and translocalization of Cdk5 in Drosophila neuronal cells. Conversion of p35 into p25 was not observed in Abeta42-triggered Drosophila neurodegeneration, suggesting that Cdk5 activation and protein translocalization can be p25-independent. Our genetic studies also showed that abl mutations repressed Abeta42-induced Cdk5 activity and neurodegeneration in Drosophila eyes. Although Abeta42 induced conversion of p35 to p25 in mammalian cells, it did not sufficiently induce Cdk5 activation when c-Abl kinase activity was suppressed. Therefore, we propose that Abl and p35/p25 cooperate in promoting Cdk5-pY15, which deregulates Cdk5 activity and subcellular localization in Abeta42-triggered neurodegeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cdk5 Antibody, clone DC17, clone DC17, Upstate®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.