Merck
The rapid increase of circulating adiponectin in neonatal calves depends on colostrum intake.
Journal of dairy science (2015-08-19)
J Kesser, M Hill, J F L Heinz, C Koch, J Rehage, J Steinhoff-Wagner, H M Hammon, B Mielenz, H Sauerwein, H Sadri
ZUSAMMENFASSUNG
Adiponectin, an adipokine, regulates metabolism and insulin sensitivity. Considering that the transplacental transfer of maternal proteins of high molecular weight is hindered in ruminants, this study tested the hypothesis that the blood concentration of adiponectin in neonatal calves largely reflects their endogenous synthesis whereby the intake of colostrum might modify the circulating concentrations. We thus characterized the adiponectin concentrations in neonatal and young calves that were fed either colostrum or formula. Three trials were performed: in trial 1, 20 calves were all fed colostrum for 3 d, and then formula until weaning. Blood samples were collected on d 0 (before colostrum feeding), and on d 1, 3, 11, 22, 34, 43, 52, 70, 90, and 108 postnatum. In trial 2, 14 calves were studied for the first 4 d of life. They were fed colostrum (n=7) or formula (n=7), and blood samples were taken right after birth and before each morning feeding on d 2, 3, and 4. In trial 3, calves born preterm (n=7) or at term received colostrum only at 24 h postnatum. Blood was sampled at birth, and before and 2 h after feeding. Additionally, allantoic fluid and blood from 4 Holstein cows undergoing cesarean section were sampled. Adiponectin was quantified by ELISA. In trial 1, the serum adiponectin concentrations recorded on d 3 were 4.7-fold higher than before colostrum intake. The distribution of the molecular weight forms of adiponectin differed before and after colostrum consumption. In trial 2, the colostrum group had consistently greater plasma adiponectin concentrations than the formula group after the first meal. In trial 3, the preterm calves tended to have lower concentrations of plasma adiponectin than the term calves at birth and before and 2 h after feeding. Furthermore, the adiponectin concentrations were substantially lower in allantoic fluid than in the sera from neonatal calves and from cows at parturition. Our results show that calves are born with very low blood concentrations of adiponectin and placental transfer of adiponectin to the bovine fetus is unlikely. In conclusion, colostrum intake is essential for the postnatal increase of circulating adiponectin in newborn calves.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-Peroxidaseantikörper (γ-kettenspezifisch), Maus, monoklonal, clone RG-96, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.