Merck
Analysis of CFTR interactome in the macromolecular complexes.
Methods in molecular biology (Clifton, N.J.) (2011-05-20)
Chunying Li, Anjaparavanda P Naren
ZUSAMMENFASSUNG
Cystic fibrosis transmembrane conductance regulator (CFTR) is a chloride channel localized primarily at the apical surface of epithelial cells lining the airway, gut, exocrine glands, etc., where it is responsible for transepithelial salt and water transport. A growing number of proteins have been reported to interact directly or indirectly with CFTR chloride channel, suggesting that CFTR might regulate the activities of other ion channels, receptors, and transporters, in addition to its role as a chloride conductor. Most interactions occur primarily between the opposing terminal tails (N or C) of CFTR and its binding partners, either directly or mediated through various PDZ domain-containing proteins. This chapter describes methods we developed to cross-link CFTR into a macromolecular complex to identify and analyze the assembly and regulation of CFTR-containing complexes in the plasma membrane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, clone MM13-4, clone MM13-4, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.