Merck
  • FRET biosensor-based kinase inhibitor screen for ERK and AKT activity reveals differential kinase dependencies for proliferation in TNBC cells.
FRET biosensor-based kinase inhibitor screen for ERK and AKT activity reveals differential kinase dependencies for proliferation in TNBC cells.
Biochemical pharmacology (2019-09-20)
Jichao He, Steven Wink, Hans de Bont, Sylvia Le Dévédec, Yinghui Zhang, Bob van de Water
ZUSAMMENFASSUNG
Enhanced expression and activity of protein kinases are critical in tumor cell proliferation and cancer progression. These various cancer-related kinases form intricate interdependent signaling networks. Evaluation of the effect of various kinase inhibitors on these networks is critical to understand kinase inhibitor efficacy in cancer therapy. The dynamic activation of some kinases can be monitored by fluorescence resonance energy transfer (FRET) biosensors with high temporal resolution. Here, we established a FRET biosensor-based high throughput imaging approach to determine ERK and AKT activity in two triple negative breast cancer (TNBC) cell lines HCC1806 and Hs578T. FRET functionality was systematically evaluated using EGF stimulation and different MEK and AKT inhibitors, respectively. Next, we assessed the effect of a kinase inhibitor library containing >350 different kinase inhibitors (KIs) on ERK and AKT kinase activity using a FRET high-throughput screening setting. Suppression of FRET-ERK activity was generally positively correlated with the proliferation phenotype against inhibitors targeting MAPK signaling in both cell lines containing FRET-ERK reporter. AKT inhibitor (AKTi) resistant HCC1806 showed decreased proliferation associated with downregulated dynamics of FRET-ERK when treated with KIs targeting protein receptor tyrosine kinase (RTK). Yet, MEK inhibitor (MEKi) resistant Hs578T showed positively correlated FRET-AKT and proliferative responses against different PI3K and AKT inhibitors. Altogether, our data demonstrate the feasibility to integrate high throughput imaging-based screening of intracellular kinase activity using FRET-based biosensors in assessing kinase specificity and possible signaling crosstalk in direct relation to therapeutic outcome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Blasticidin S -hydrochlorid