Merck
  • Partitioning of MLX-Family Transcription Factors to Lipid Droplets Regulates Metabolic Gene Expression.
Partitioning of MLX-Family Transcription Factors to Lipid Droplets Regulates Metabolic Gene Expression.
Molecular cell (2020-02-06)
Niklas Mejhert, Leena Kuruvilla, Katlyn R Gabriel, Shane D Elliott, Marie-Aude Guie, Huajin Wang, Zon Weng Lai, Elizabeth A Lane, Romain Christiano, Nika N Danial, Robert V Farese, Tobias C Walther
ZUSAMMENFASSUNG
Lipid droplets (LDs) store lipids for energy and are central to cellular lipid homeostasis. The mechanisms coordinating lipid storage in LDs with cellular metabolism are unclear but relevant to obesity-related diseases. Here we utilized genome-wide screening to identify genes that modulate lipid storage in macrophages, a cell type involved in metabolic diseases. Among ∼550 identified screen hits is MLX, a basic helix-loop-helix leucine-zipper transcription factor that regulates metabolic processes. We show that MLX and glucose-sensing family members MLXIP/MondoA and MLXIPL/ChREBP bind LDs via C-terminal amphipathic helices. When LDs accumulate in cells, these transcription factors bind to LDs, reducing their availability for transcriptional activity and attenuating the response to glucose. Conversely, the absence of LDs results in hyperactivation of MLX target genes. Our findings uncover a paradigm for a lipid storage response in which binding of MLX transcription factors to LD surfaces adjusts the expression of metabolic genes to lipid storage levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Millipore
Benzonase® Nuklease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
OptiPrep Dichtegradientenmedium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Insulin aus Rinderpankreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Dexamethason – wasserlöslich, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
3,3′-Diiod-L-thyronin (T2), 98% (CP)