Merck
  • Granzyme B Produced by Natural Killer Cells Enhances Inflammatory Response and Contributes to the Immunopathology of Cutaneous Leishmaniasis.
Granzyme B Produced by Natural Killer Cells Enhances Inflammatory Response and Contributes to the Immunopathology of Cutaneous Leishmaniasis.
The Journal of infectious diseases (2019-11-22)
Taís M Campos, Fernanda O Novais, Maíra Saldanha, Rúbia Costa, Morgana Lordelo, Daniela Celestino, Camilla Sampaio, Natália Tavares, Sérgio Arruda, Paulo Machado, Cláudia Brodskyn, Phillip Scott, Edgar M Carvalho, Lucas P Carvalho
ZUSAMMENFASSUNG
Skin lesions from patients infected with Leishmania braziliensis has been associated with inflammation induced by cytotoxic CD8+ T cells. In addition, CD8+ T cell-mediated cytotoxicity has not been linked to parasite killing. Meanwhile, the cytotoxic role played by natural killer (NK) cells in cutaneous leishmaniasis (CL) remains poorly understood. In this study, we observed higher frequencies of NK cells in the peripheral blood of CL patients compared with healthy subjects, and that NK cells expressed more interferon-γ, tumor necrosis factor (TNF), granzyme B, and perforin than CD8+ T cells. We also found that most of the cytotoxic activity in CL lesions was triggered by NK cells, and that the high levels of granzyme B produced in CL lesions was associated with larger lesion size. Furthermore, an in vitro blockade of granzyme B was observed to decrease TNF production. Our data, taken together, suggest an important role by NK cells in inducing inflammation in CL, thereby contributing to disease immunopathology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dichlorisocumarin, serine protease inhibitor
Preise und Verfügbarkeit sind derzeit nicht verfügbar.