Merck
Creatine uptake regulates CD8 T cell antitumor immunity.
The Journal of experimental medicine (2019-10-20)
Stefano Di Biase, Xiaoya Ma, Xi Wang, Jiaji Yu, Yu-Chen Wang, Drake J Smith, Yang Zhou, Zhe Li, Yu Jeong Kim, Nicole Clarke, Angela To, Lili Yang
ZUSAMMENFASSUNG
T cells demand massive energy to combat cancer; however, the metabolic regulators controlling antitumor T cell immunity have just begun to be unveiled. When studying nutrient usage of tumor-infiltrating immune cells in mice, we detected a sharp increase of the expression of a CrT (Slc6a8) gene, which encodes a surface transporter controlling the uptake of creatine into a cell. Using CrT knockout mice, we showed that creatine uptake deficiency severely impaired antitumor T cell immunity. Supplementing creatine to WT mice significantly suppressed tumor growth in multiple mouse tumor models, and the combination of creatine supplementation with a PD-1/PD-L1 blockade treatment showed synergistic tumor suppression efficacy. We further demonstrated that creatine acts as a "molecular battery" conserving bioenergy to power T cell activities. Therefore, our results have identified creatine as an important metabolic regulator controlling antitumor T cell immunity, underscoring the potential of creatine supplementation to improve T cell-based cancer immunotherapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fetales Kälberserum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Adenosin-5′-triphosphat Dinatriumsalz Hydrat, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Creatin Monohydrat, ≥98%
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder