- Growth and maintenance of Vero cell lines.
Growth and maintenance of Vero cell lines.
Current protocols in microbiology (2008-11-20)
Nicole C Ammerman, Magda Beier-Sexton, Abdu F Azad
PMID19016439
ZUSAMMENFASSUNG
Vero cells are derived from the kidney of an African green monkey, and are one of the more commonly used mammalian continuous cell lines in microbiology and molecular and cell biology research. This unit includes protocols for the growth and maintenance of Vero cell lines in a research laboratory setting.