Merck
  • Distinct roles of srGAP3-Rac1 in the initiation and maintenance phases of neuropathic pain induced by paclitaxel.
Distinct roles of srGAP3-Rac1 in the initiation and maintenance phases of neuropathic pain induced by paclitaxel.
The Journal of physiology (2020-04-03)
Zihang Chen, Subo Zhang, Bilin Nie, Jingxiu Huang, Zhixiao Han, Xiaodi Chen, Xiaohui Bai, Handong Ouyang
ZUSAMMENFASSUNG
Spinal cord dorsal horn srGAP3 (slit-robo GTPase activating protein 3) increases in the initiation phase of neuropathic pain and decreases in the maintenance phase. However, Rac1 activity, which can be reduced by srGAP3, decreases in the initiation phase and increases in the maintenance phase. The increased srGAP3 in the initiation phase promotes new immature dendritic spines instigating neuropathic pain. Decreased srGAP3 in the maintenance phase enhances Rac1 activity facilitating maturation of dendritic spines and the persistence of neuropathic pain. SrGAP3 small interfering RNA can ameliorate neuropathic pain only when administrated in the initiation phase. The Rac1 inhibitor can ameliorate neuropathic pain only when administrated in the maintenance phase. Combined targeting of srGAP3 in the initiation phase and Rac1 in the maintenance phase can produce optimal analgesic efficacy. Neuropathic pain includes an initiation phase and maintenance phase, each with different pathophysiological processes. Understanding the synaptic plasticity and molecular events in these two phases is relevant to exploring precise treatment strategies for neuropathic pain. In the present study, we show that dendritic spine density increases in the spinal dorsal horn in the initiation phase of neuropathic pain induced by paclitaxel and that the spine maturity ratio increases in the maintenance phase. Increased srGAP3 (slit-robo GTPase activating protein 3) facilitates dendritic spine sprouting in the initiation phase. In the maintenance phase, srGAP3 decreases to upregulate Rac1 activity, which facilitates actin polymerization and dendritic spine maturation and thus the persistence of neuropathic pain. Knockdown of srGAP3 in the initiation phase or inhibition of Rac1 in the maintenance phase attenuates neuropathic pain. Combined intervention of srGAP3 in the initiation phase, and Rac1 in the maintenance phase shows better analgesic efficacy against neuropathic pain. The present study demonstrates the role of srGAP3-Rac1 in dendritic spine plasticity in the two phases of neuropathic pain and, accordingly, provides treatment strategies for different phases of neuropathic pain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Protease- und Phosphatasehemmer-Cocktail
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, clone C4, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, Magnetic bead Conjugate, purified antibody, clone 23A8, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.