Merck
  • Therapeutic potential of miR-21 regulation by human peripheral blood derived-small extracellular vesicles in myocardial infarction.
Therapeutic potential of miR-21 regulation by human peripheral blood derived-small extracellular vesicles in myocardial infarction.
Clinical science (London, England : 1979) (2020-04-17)
Ji-Young Kang, Hyoeun Kim, Dasom Mun, Nuri Yun, Boyoung Joung
ZUSAMMENFASSUNG
Small extracellular vesicles (sEVs) as natural membranous vesicles are on the frontiers of nanomedical research, due to their ability to deliver therapeutic molecules such as microRNAs (miRNAs). The miRNA-21 (miR-21) is thought to be involved in the initiation and development of myocardial infarction (MI). Here, we examined whether miR-21 regulation using human peripheral blood-derived sEVs (PB-sEVs) could serve as a potential therapeutic strategy for MI. First, we examined miR-21 levels in hypoxic conditions and validated the ability of PB-sEVs to serve as a potential delivery system for miRNAs. Further, bioinformatics analysis and luciferase assay were performed to identify target genes of miR-21 mechanistically. Among numerous target pathways, we focused on nitrogen metabolism, which remains relatively unexplored compared with other possible miR-21-mediated pathways; hence, we aimed to determine novel target genes of miR-21 related to nitrogen metabolism. In hypoxic conditions, the expression of miR-21 was significantly up-regulated and correlated with nitric oxide synthase 3 (NOS3) levels, which in turn influences cardiac function. The down-regulation of miR-21 expression by PB-sEVs loaded with anti-miR-21 significantly improved survival rates, consistent with the augmentation of cardiac function. However, the up-regulation of miR-21 expression by PB-sEVs loaded with miR-21 reversed these effects. Mechanistically, miR-21 targeted and down-regulated the mRNA and protein expression of striatin (STRN), which could regulate NOS3 expression. In conclusion, we identified a novel therapeutic strategy to improve cardiac function by regulating the expression of miR-21 with PB-sEVs as an miR-21 or anti-miR-21 delivery vehicle and confirmed the miR-21-associated nitrogen metabolic disorders in MI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronektin-Rinderplasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Immobilon®-P PVDF-Membran, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Claycomb-Medium, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
PKH26 Linker-Kit für rot fluoreszierende Zellen für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(±)-Norepinephrin -(+)-bitartratsalz
Preise und Verfügbarkeit sind derzeit nicht verfügbar.