Merck
Electrogenetic cellular insulin release for real-time glycemic control in type 1 diabetic mice.
Science (New York, N.Y.) (2020-05-30)
Krzysztof Krawczyk, Shuai Xue, Peter Buchmann, Ghislaine Charpin-El-Hamri, Pratik Saxena, Marie-Didiée Hussherr, Jiawei Shao, Haifeng Ye, Mingqi Xie, Martin Fussenegger
ZUSAMMENFASSUNG
Sophisticated devices for remote-controlled medical interventions require an electrogenetic interface that uses digital electronic input to directly program cellular behavior. We present a cofactor-free bioelectronic interface that directly links wireless-powered electrical stimulation of human cells to either synthetic promoter-driven transgene expression or rapid secretion of constitutively expressed protein therapeutics from vesicular stores. Electrogenetic control was achieved by coupling ectopic expression of the L-type voltage-gated channel CaV1.2 and the inwardly rectifying potassium channel Kir2.1 to the desired output through endogenous calcium signaling. Focusing on type 1 diabetes, we engineered electrosensitive human β cells (Electroβ cells). Wireless electrical stimulation of Electroβ cells inside a custom-built bioelectronic device provided real-time control of vesicular insulin release; insulin levels peaked within 10 minutes. When subcutaneously implanted, this electrotriggered vesicular release system restored normoglycemia in type 1 diabetic mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Resazurin, powder, BioReagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluidinblau, suitable for microscopy, 8.74% (ZN (THEORY))
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Alloxan Monohydrat, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.