Merck
  • Comparison of acute treatment with delayed-onset versus rapid-acting antidepressants on effort-related choice behaviour.
Comparison of acute treatment with delayed-onset versus rapid-acting antidepressants on effort-related choice behaviour.
Psychopharmacology (2020-05-22)
Simonas Griesius, Jack R Mellor, Emma Sj Robinson
ZUSAMMENFASSUNG
Reward-related impairments are common in major depressive disorder (MDD) and may contribute to the loss of interest in pleasurable activities. A novel approach to studying reward-related decision-making are effort-based tasks; however, direct comparisons between delayed-onset and rapid-acting antidepressants (ADs) have not yet been carried out. To investigate the effects of conventional delayed-onset ADs versus rapid-acting ADs, ketamine and scopolamine, on effort-related choice behaviour. Female Lister hooded rats were trained in an operant effort for reward task (EfRT) where animals choose between working for a high value-high effort reward and consuming low value-low effort chow. Using a within-subject study design, animals were then tested following acute treatment with different monoaminergic ADs, and the rapid-acting ADs ketamine or scopolamine. Consistent with previous findings, we found choice behaviour was sensitive to dopaminergic manipulations. We observed that pre-feeding altered choice behaviour and that the use of high or low value reward differentially affected behaviour. Monoamine re-uptake inhibitors and rapid-acting ADs resulted in similar, general patterns of reduced motivation without any evidence for specific effects, and we did not observe any clear differences between these classes of antidepressant. Motivational changes induced by dopaminergic manipulations and pre-feeding differentially affect effort choice behaviour. However, both conventional delayed-onset ADs and ketamine and scopolamine appear to have detrimental effects on motivation in this task at the higher doses tested without any evidence of specificity for effort-related choice behaviour, in contrast to their specificity in tasks which look at more cognitive aspects of reward processing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tetrabenazin, ≥98% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.