Merck
  • Lipophilic Prodrug of Methotrexate in the Membrane of Liposomes Promotes Their Uptake by Human Blood Phagocytes.
Lipophilic Prodrug of Methotrexate in the Membrane of Liposomes Promotes Their Uptake by Human Blood Phagocytes.
Acta naturae (2020-06-02)
D S Tretiakova, S V Khaidukov, A A Babayants, I S Frolova, O N Shcheglovitova, N R Onishchenko, E L Vodovozova
ZUSAMMENFASSUNG
Previously, we showed that incorporation of methotrexate (MTX) in the form of a lipophilic prodrug (MTXDG) in 100-nm lipid bilayer liposomes of egg phosphatidylcholine can allow one to reduce toxicity and improve the antitumor efficiency of MTX in a mouse model of T-cell leukemic lymphoma. However, in our hemocompatibility tests in vitro, MTX liposomes caused complement (C) activation, obviously due to binding on the liposome surface and fragmentation of the C3 complement factor. In this work, we studied the interactions of MTX liposomes carrying stabilizing molecules phosphatidylinositol (PI), ganglioside GM1, or a lipid conjugate of N-carboxymethylated oligoglycine (CMG) in the bilayer with subpopulations of human blood leukocytes. Liposomes labeled with BODIPY-phosphatidylcholine were incubated with whole blood (30 min and 1 h, 37°C), blood cells were lysed with a hypotonic buffer, and the fluorescence of the liposomes bound but not internalized by the leukocytes was quenched by crystal violet. Cell suspensions were analyzed by flow cytometry. Incorporation of MTXDG dramatically enhanced the phagocytosis of liposomes of any composition by monocytes. Neutrophils consumed much less of the liposomes. Lymphocytes did not accumulate liposomes. The introduction of PI into MTX liposomes practically did not affect the specific consumption of liposomes by monocytes, while CMG was likely to increase the consumption rate regardless of the presence of MTXDG. The GM1 ganglioside presumably shielded MTX liposomes from phagocytosis by one of the monocyte populations and increased the efficiency of monocyte uptake by another population, probably one expressing C3b-binding receptors (C3b was detected on liposomes after incubation with blood plasma). MTX liposomes were shown to have different effects on TNF-α production by activated leukocytes, depending on the structure of the stabilizing molecule.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Phosphonohexansäure, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.