Merck
  • Effects of oocyte vitrification on the behaviors and physiological indexes of aged first filial generation mice.
Effects of oocyte vitrification on the behaviors and physiological indexes of aged first filial generation mice.
Cryobiology (2020-07-01)
Ying Huo, Qingyuan Qin, Lu Zhang, Ying Kuo, Haiyan Wang, Liying Yan, Rong Li, Xiaowei Zhang, Jie Yan, Jie Qiao
ZUSAMMENFASSUNG
To evaluate the long-term effects of oocyte cryopreservation on the health of the first filial generation (F1), we used B6D2F1 mice for oocyte collection, in vitro fertilization, and breeding. The female F1 mice born from the offspring of fresh mature oocytes (control group) and from the offspring of vitrified oocytes with traditional vitrification medium (VM group) and new improved vitrification medium (2P10E7D group) were maintained until 14-15 months of age for behavioral tests and 16-17 months of age for physiological analyses. Behavioral indexes, including anxiety-like status, discrimination ability, learning and memory ability, were investigated. Physiological indexes including body weight, body fat, heart rate, blood pressure, and blood lipids were also analyzed. In our results, the behavioral indexes, body weight, body fat, heart rate, blood pressure, total cholesterol (TC), high-density lipoprotein cholesterol (HDL), and low-density lipoprotein cholesterol (LDL) did not show significant differences among the three groups. However, the triglyceride (TG) level of the VM group was higher than that of the 2P10E7D group. Moreover, compared with the control group, both the VM group and the 2P10E7D group showed greatly increased diastolic blood pressure. This study is the first to report that oocyte vitrification might affect metabolic physiological indexes via transgenerational inheritance rather than behaviors related to anxiety-like status and cognitive ability. Furthermore, different vitrification media might have differential transgenerational effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.