Merck
  • Bone marrow mesenchymal stem cells upregulate PI3K/AKT pathway and down-regulate NF-κB pathway by secreting glial cell-derived neurotrophic factors to regulate microglial polarization and alleviate deafferentation pain in rats.
Bone marrow mesenchymal stem cells upregulate PI3K/AKT pathway and down-regulate NF-κB pathway by secreting glial cell-derived neurotrophic factors to regulate microglial polarization and alleviate deafferentation pain in rats.
Neurobiology of disease (2020-05-20)
Zhenzhong Zhong, Ao Chen, Zhiqiang Fa, Zhiquan Ding, Linglong Xiao, Guiwei Wu, Qinghua Wang, Run Zhang
ZUSAMMENFASSUNG
Deafferentation pain (DP), a typical neuropathic pain, occurs due to peripheral or central sensory nerve injury, which causes abnormal discharge of the upstream neurons or C fibers. Current treatment methods for DP have multiple side effects. Bone marrow mesenchymal stem cells (BMSC) have been used to treat neuropathic pain because of their ability to regulate neuroinflammation. Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic mediator that exerts neuroprotective effects in neurological diseases. In this study, we investigated whether DP could be alleviated by BMSCs and the underlying mechanism. In vitro study, microglia was stimulated by lipopolysaccharide and then co-cultured with BMSC, GDNF or siRNA GDNF-BMSC. In vivo study, BMSC or siRNA GDNF-BMSC was transplanted intramedullarily on the 21st day after DP surgery. The expression of inflammatory-related factors were detected by RT-PCR and ELISA, RT-PCR,flow cytometry and immunofluorescence staining were performed to detect the expression of microglial surface markers, and Western blot was used to detect the expression levels of p-NF-kb, pPI3K, and pAKT. The pain-related behavioral changes were detected 7 days after transplantation. ELISA and RT-PCR results showed that the production of inflammatory cytokines in lipopolysaccharide-stimulated microglia and DP model plasma was downregulated, while anti-inflammatory mediators were upregulated significantly following pretreatment with BMSCs or GDNF. Flow cytometry, immunofluorescence staining, and RT-PCR results showed that BMSCs inhibited the microglial M1 phenotype and promoted the M2 phenotype by secreting GDNF. Furthermore, modulation functions of BMSCs involve inhibiting NF-κB while promoting PI3K /AKT signaling pathway activation. We found that our in vivo DP model was completely deafferent and BMSC administration clearly alleviated symptoms of DP. This function was also, at least partly, achieved by GDNF. The present studies demonstrate that BMSC can inhibit neuroinflammation by transforming microglial destructive M1 phenotype into regenerative M2 phenotype, and thus alleviate DP,likely by suppressing the NF-κB signaling pathway while promoting the PI3K/AKT signaling pathway activation through producing GDNF. The present findings are in support of the potential therapeutic application of BMSCs and the pharmaceutical application of GDNF for DP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human GDNF
Preise und Verfügbarkeit sind derzeit nicht verfügbar.