Merck
  • Oxidized low-density lipoprotein inhibits the degradation of cyclophilin A via the lysosome in vascular smooth muscle cells.
Oxidized low-density lipoprotein inhibits the degradation of cyclophilin A via the lysosome in vascular smooth muscle cells.
American journal of translational research (2020-08-11)
Zizhuo Su, Maohuan Lin, Yinqing Su, Jiajie Li, Rongjie Lin, Maoxiong Wu, Xiaoyu Wang, Lili Huang, Yuyang Chen, Xiaorong Shu, Shumin Liang, Haifeng Zhang, Tucheng Huang, Ruqiong Nie, Jingfeng Wang, Shuanglun Xie
ZUSAMMENFASSUNG
Cyclophilin A (CyPA) plays an important role in the progression of atherosclerosis. Additionally, it has been reported that lysosomal function is markedly impaired in atherosclerosis induced by oxidized low-density lipoprotein (ox-LDL). As the CyPA degradation pathway remains to be elucidated, we aimed to uncover the role of lysosomes and ox-LDL in the degradation of CyPA. We exploited RNA interference (RNAi) in combination with either the lysosomal inhibitor chloroquine (CQ) or the proteasomal inhibitor MG-132 to examine CyPA turnover. We also investigated the role of ox-LDL in lysosomal function and the CyPA degradation pathway and determined whether CyPA interacts with the selective autophagy adaptor p62. CQ markedly reversed the CyPA downregulation induced by RNAi and increased intracellular levels of LC3 and p62. MG-132 significantly suppressed polyubiquitinated protein degradation but did not inhibit RNAi-induced CyPA downregulation. Additionally, neither CQ nor MG-132 influenced the gene-silencing efficiency of CyPA siRNA. Moreover, ox-LDL induced cytosolic accumulation of p62 was inconsistent with increased expression of LC3-II. Meanwhile, ox-LDL inhibited RNAi-induced downregulation of CyPA. Immunofluorescence indicated colocalization of endogenous CyPA with ubiquitin and with p62 in response to CQ treatment, and co-immunoprecipitation analysis confirmed interaction between CyPA and p62. CyPA is degraded by a lysosome-dependent pathway that may involve p62-mediated selective autophagy. Furthermore, ox-LDL modulates the degradation of CyPA via its inhibitory role in lysosomes, contributing to increased expression of CyPA in atherosclerotic plaques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-p62/SQSTM1 in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.