Merck
  • UNC-129 regulates the balance between UNC-40 dependent and independent UNC-5 signaling pathways.
UNC-129 regulates the balance between UNC-40 dependent and independent UNC-5 signaling pathways.
Nature neuroscience (2009-01-27)
Lesley T MacNeil, W Rod Hardy, Tony Pawson, Jeffrey L Wrana, Joseph G Culotti
ZUSAMMENFASSUNG
The UNC-5 receptor mediates axon repulsion from UNC-6/netrin through UNC-40 dependent (UNC-5 + UNC-40) and independent (UNC-5 alone) signaling pathways. It has been shown that UNC-40-dependent signaling is required for long-range repulsion of UNC-6/netrin; however, the mechanisms used to regulate distinct UNC-5 signaling pathways are poorly understood. We found that the C. elegans transforming growth factor beta (TGF-beta) family ligand UNC-129, graded opposite to UNC-6/netrin, functions independent of the canonical TGF-beta receptors to regulate UNC-5 cellular responses. Our observations indicates that UNC-129 facilitates long-range repulsive guidance of UNC-6 by enhancing UNC-5 + UNC-40 signaling at the expense of UNC-5 alone signaling through interaction with the UNC-5 receptor. This increases the set point sensitivity of growth cones to UNC-6/netrin as they simultaneously migrated up the UNC-129 gradient and down the UNC-6 gradient. Similar regulatory interactions between oppositely graded extracellular cues may be a common theme in guided cell and axon migrations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.1 % (w/v) in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Poly-L-Lysin -Lösung, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.