Merck
  • Visible light photo-oxidation of model pollutants using CaCu3Ti4O12: an experimental and theoretical study of optical properties, electronic structure, and selectivity.
Visible light photo-oxidation of model pollutants using CaCu3Ti4O12: an experimental and theoretical study of optical properties, electronic structure, and selectivity.
Journal of the American Chemical Society (2010-12-17)
Joanna H Clark, Matthew S Dyer, Robert G Palgrave, Christopher P Ireland, James R Darwent, John B Claridge, Matthew J Rosseinsky
ZUSAMMENFASSUNG
Charge transfer between metal ions occupying distinct crystallographic sublattices in an ordered material is a strategy to confer visible light absorption on complex oxides to generate potentially catalytically active electron and hole charge carriers. CaCu3Ti4O12 has distinct octahedral Ti4+ and square planar Cu2+ sites and is thus a candidate material for this approach. The sol−gel synthesis of high surface area CaCu3Ti4O12 and investigation of its optical absorption and photocatalytic reactivity with model pollutants are reported. Two gaps of 2.21 and 1.39 eV are observed in the visible region. These absorptions are explained by LSDA+U electronic structure calculations, including electron correlation on the Cu sites, as arising from transitions from a Cu-hybridized O 2p-derived valence band to localized empty states on Cu (attributed to the isolation of CuO4 units within the structure of CaCu3Ti4O12) and to a Ti-based conduction band. The resulting charge carriers produce selective visible light photodegradation of 4-chlorophenol (monitored by mass spectrometry) by Pt-loaded CaCu3Ti4O12 which is attributed to the chemical nature of the photogenerated charge carriers and has a quantum yield comparable with commercial visible light photocatalysts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Supelco
Ethylenglycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
L-(+)-Tartarsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Oxalsäure, purified grade, 99.999% trace metals basis
Supelco
Citronensäure, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxalsäure, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Ammoniumhydroxid -Lösung, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Ethylenglycol, analytical standard
Sigma-Aldrich
Titan(IV)-bis(ammoniumlactato)dihydroxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
L-(+)-Tartarsäure, ≥99.5%
Sigma-Aldrich
L-(+)-Tartarsäure, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethylenglycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-(+)-Tartarsäure, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Supelco
Zitronensäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-(+)-Tartarsäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartarsäure, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
L-(+)-Tartarsäure, BioXtra
Supelco
L-(+)-Tartarsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland