Merck
Hierarchical clustering of monoclonal antibody reactivity patterns in nonhuman species.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2009-07-30)
Juan Pablo Pratt, Qing Zeng, Dino Ravnic, Harold Huss, James Rawn, Steven J Mentzer
ZUSAMMENFASSUNG
Monoclonal antibodies (Mab) are an important resource for defining molecular expression and probing molecular function. The characterization of Mab reactivity patterns, however, can be costly and inefficient in nonhuman experimental systems. To develop a computational approach to the pattern analysis of Mab reactivity, we analyzed a panel of 128 Mab recognizing sheep antigens. Quantitative single parameter flow cytometry histograms were obtained from five cell types isolated from normal animals. The resulting 640 histograms were smoothed using a Gaussian kernel over a range of bandwidths. Histogram features were selected by SiZer--an analytic tool that identifies statistically significant features. The extracted histogram features were compared and grouped using hierarchical clustering. The validity of the clustering was indicated by the accurate pairing of externally verified molecular reactivity. We conclude that our computational algorithm is a potentially useful tool for both Mab classification and molecular taxonomy in nonhuman experimental systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
RPMI-1640-Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Polyethylenglycol-Lösung, Hybri-Max, 50 % (w/v), average mol wt 1,450, 0.2 μm filtered, BioReagent, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
RPMI-1640-Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Polyethylenglykol -Lösung, 40 % (w/w) in H2O, average mol wt 8,000
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
RPMI-1640-Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
RPMI-1640-Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Endothelial cell growth supplement from bovine pituitary, ECGS, suitable for cell culture, BioReagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Polyethylenglykol -Lösung, BioUltra, Molecular Biology, 1,000, ~50% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
RPMI-1640-Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
RPMI-1640-Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Polyethylenglycol-/Dimethylsulfoxid-Lösung, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Streptomycin -Lösung, ~1 mg/mL in 1 mM EDTA, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.