Merck
  • Bone and Mineral Metabolism in Children with Nephropathic Cystinosis Compared with other CKD Entities.
Bone and Mineral Metabolism in Children with Nephropathic Cystinosis Compared with other CKD Entities.
The Journal of clinical endocrinology and metabolism (2020-05-16)
Annika Ewert, Maren Leifheit-Nestler, Katharina Hohenfellner, Anja Büscher, Markus J Kemper, Jun Oh, Heiko Billing, Julia Thumfart, Gabriele Stangl, Anja C Baur, Michael Föller, Martina Feger, Lutz T Weber, Birgit Acham-Roschitz, Klaus Arbeiter, Burkhard Tönshoff, Miroslav Zivicnjak, Dieter Haffner
ZUSAMMENFASSUNG
Children with nephropathic cystinosis (NC) show persistent hypophosphatemia, due to Fanconi syndrome, as well as mineral and bone disorders related to chronic kidney disease (CKD); however, systematic analyses are lacking. To compare biochemical parameters of bone and mineral metabolism between children with NC and controls across all stages of CKD. Cross-sectional multicenter study. Hospital clinics. Forty-nine children with NC, 80 CKD controls of the same age and CKD stage. Fibroblast growth factor 23 (FGF23), soluble Klotho, bone alkaline phosphatase (BAP), tartrate-resistant acid phosphatase 5b (TRAP5b), sclerostin, osteoprotegerin (OPG), biochemical parameters related to mineral metabolism, and skeletal comorbidity. Despite Fanconi syndrome medication, NC patients showed an 11-fold increased risk of short stature, bone deformities, and/or requirement for skeletal surgery compared with CKD controls. This was associated with a higher frequency of risk factors such as hypophosphatemia, hypocalcemia, low parathyroid hormone (PTH), metabolic acidosis, and a specific CKD stage-dependent pattern of bone marker alterations. Pretransplant NC patients in mild to moderate CKD showed a delayed increase or lacked an increase in FGF23 and sclerostin, and increased BAP, TRAP5b, and OPG concentrations compared with CKD controls. Post-transplant, BAP and OPG returned to normal, TRAP5b further increased, whereas FGF23 and PTH were less elevated compared with CKD controls and associated with higher serum phosphate. Patients with NC show more severe skeletal comorbidity associated with distinct CKD stage-dependent alterations of bone metabolism than CKD controls, suggesting impaired mineralization and increased bone resorption, which is only partially normalized after renal transplantation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,4-Bis-acryloyl-piperazin, BioXtra, suitable for electrophoresis, ≥99.0% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.