Merck
  • Dental Adhesive Interfaces Reinforced with Magnetic Nanoparticles: Evaluation and Modeling with Micro-CT versus Optical Microscopy.
Dental Adhesive Interfaces Reinforced with Magnetic Nanoparticles: Evaluation and Modeling with Micro-CT versus Optical Microscopy.
Materials (Basel, Switzerland) (2020-09-10)
Cristian Zaharia, Virgil-Florin Duma, Cosmin Sinescu, Vlad Socoliuc, Izabell Craciunescu, Rodica Paula Turcu, Catalin Nicolae Marin, Anca Tudor, Mihai Rominu, Meda-Lavinia Negrutiu
ZUSAMMENFASSUNG
Dental adhesives are used in a wide range of applications, including to place direct composite restorations in frontal or posterior teeth. One of the most frequent causes for the failure of composite resin restorations is microleakages. The first aim of this work is to introduce a new type of self-etched dental adhesive doped with magnetic nanoparticles (MPs) synthetized in the laboratory. The scope is to produce adhesives with a minimized width/thickness to decrease the risk of microleakages. The second aim is to assess the width/thickness of the adhesive layer in all the characteristic areas of the teeth using both the less precise but most common optical microscopy and the more accurate and volumetric micro-Computed Tomography (CT) investigations. Twenty extracted teeth have been divided into four groups: Group 1 includes 'blank' samples with adhesives that are not doped with MPs; Group 2 includes samples with adhesives doped with MPs; Groups 3 and 4 include samples with adhesives doped with MPs that are subjected to an active magnetic field for 5 and 10 min, respectively. Microscopy investigations followed by micro-CT and EDAX are performed on the adhesive. While a rather good agreement is obtained between the microscopy and micro-CT results, the capability of the latter to offer a full volumetric reconstruction of the layer is exploited to analyze the adhesion of the four considered dental materials. Thus, from micro-CT results the graphs of the surface areas as functions of the adhesive layer width are modeled mathematically, as well as the volume of sealants, for each of the four groups. To our knowledge, it is the first time that such a methodology is used. Characteristic parameters are extracted and the ascertainment of the optimal parameter that should be utilized for such assessments is discussed. The study demonstrates the adhesion improvement produced for Groups 3 and 4, where MPs are used. It also concludes that the magnetic field should be applied to the adhesive material for the longest possible exposure time (with a trade-off with the clinical duration of the treatment).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.