Merck
  • Mitochondrial Transfer by Human Mesenchymal Stromal Cells Ameliorates Hepatocyte Lipid Load in a Mouse Model of NASH.
Mitochondrial Transfer by Human Mesenchymal Stromal Cells Ameliorates Hepatocyte Lipid Load in a Mouse Model of NASH.
Biomedicines (2020-09-18)
Mei-Ju Hsu, Isabel Karkossa, Ingo Schäfer, Madlen Christ, Hagen Kühne, Kristin Schubert, Ulrike E Rolle-Kampczyk, Stefan Kalkhof, Sandra Nickel, Peter Seibel, Martin von Bergen, Bruno Christ
ZUSAMMENFASSUNG
Mesenchymal stromal cell (MSC) transplantation ameliorated hepatic lipid load; tissue inflammation; and fibrosis in rodent animal models of non-alcoholic steatohepatitis (NASH) by as yet largely unknown mechanism(s). In a mouse model of NASH; we transplanted bone marrow-derived MSCs into the livers; which were analyzed one week thereafter. Combined metabolomic and proteomic data were applied to weighted gene correlation network analysis (WGCNA) and subsequent identification of key drivers. Livers were analyzed histologically and biochemically. The mechanisms of MSC action on hepatocyte lipid accumulation were studied in co-cultures of hepatocytes and MSCs by quantitative image analysis and immunocytochemistry. WGCNA and key driver analysis revealed that NASH caused the impairment of central carbon; amino acid; and lipid metabolism associated with mitochondrial and peroxisomal dysfunction; which was reversed by MSC treatment. MSC improved hepatic lipid metabolism and tissue homeostasis. In co-cultures of hepatocytes and MSCs; the decrease of lipid load was associated with the transfer of mitochondria from the MSCs to the hepatocytes via tunneling nanotubes (TNTs). Hence; MSCs may ameliorate lipid load and tissue perturbance by the donation of mitochondria to the hepatocytes. Thereby; they may provide oxidative capacity for lipid breakdown and thus promote recovery from NASH-induced metabolic impairment and tissue injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Mitochondrien-Antikörper, Oberfläche intakter Mitochondrien, Klon 113-1, clone 113-1, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Vinculin-Antikörper, Klon V284, clone V284, Upstate®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.