Merck
Nogo-B is associated with cytoskeletal structures in human monocyte-derived macrophages.
BMC research notes (2011-01-18)
Kathrin Schanda, Martin Hermann, Nadia Stefanova, Viktoria Gredler, Christine Bandtlow, Markus Reindl
ZUSAMMENFASSUNG
The reticulon Nogo-B participates in cellular and immunological processes in murine macrophages. Since leukocytes are an essential part of the immune system in health and disease, we decided to investigate the expression of Nogo-A, Nogo-B and Nogo-C in different human immune cell subpopulations. Furthermore, we analyzed the localization of Nogo-B in human monocyte-derived macrophages by indirect immunofluorescence stainings to gain further insight into its possible function. We describe an association of Nogo-B with cytoskeletal structures and the base of filopodia, but not with focal or podosomal adhesion sites of monocyte-derived macrophages. Nogo-B positive structures are partially co-localized with RhoA staining and Rac1 positive membrane ruffles. Furthermore, Nogo-B is associated with the tubulin network, but not accumulated in the Golgi region. Although Nogo-B is present in the endoplasmic reticulum, it can also be translocated to large cell protrusions or the trailing end of migratory cells, where it is homogenously distributed. Two different Nogo-B staining patterns can be distinguished in macrophages: firstly we observed ER-independent Nogo-B localization in cell protrusions and at the trailing end of migrating cells. Secondly, the localization of Nogo-B in actin/RhoA/Rac1 positive regions supports an influence on cytoskeletal organization. To our knowledge this is the first report on Nogo-B expression at the base of filopodia, thus providing further insight into the distribution of this protein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paraformaldehyd, powder, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Paraformaldehyd, prilled, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
CHAPS-Hydrat, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Paraformaldehyd, reagent grade, crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Paraformaldehyd, meets analytical specification of DAC, 95.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
CHAPS-Hydrat, BioReagent, suitable for electrophoresis, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
CHAPS-Hydrat, BioXtra, ≥98% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.