Merck
Novel Role of HAX-1 in Neurons Protection After Spinal Cord Injury Involvement of IRE-1.
Neurochemical research (2020-07-19)
Jiajia Chen, Saishuai Yang, Chunshuai Wu, Zhiming Cui, Yangyang Wan, Guanhua Xu, Guofeng Bao, Jinlong Zhang, Chu Chen, Dianwen Song
ZUSAMMENFASSUNG
Spinal cord injury (SCI) is one of the diseases with high probability of causing disability in human beings, and there is no reliable treatment at present. Neuronal apoptosis is a vital component of secondary injury and plays a critical role in the development of neurological dysfunction after spinal cord injury. In this study, we found that the expression and distribution of HAX-1 in neurons increased 1 day after SCI. PC12 cells overexpressing HAX-1 showed decreased apoptosis and PC12 cells are more likely to undergo apoptosis after down-regulating HAX-1, which was confirmed via TUNEL experiments. We found GRP94 showed the same trend as HAX-1 in expression and interacted with HAX-1 and IRE-1 in both spinal cord tissue and PC12 cells, and this interaction seems to be enhanced after SCI. When the expression of HAX-1 was up-regulated, GRP94 also increased, but IRE-1 did not change at all. Further studies showed that overexpression of HAX-1 decreased the expression of pIRE-1, rather than IRE-1, and downstream proteins of the IRE signaling pathway (Caspase12, pJNK and CHOP) were significantly reduced, and vice versa. In animals treated with HAX-1 expressing adenovirus there are more neuronal cells remaining in the damaged spinal cord tissue, and hindlimb motor function of rats was significantly improved. So, we speculate that HAX-1 might play a role in protecting neurons from apoptosis after SCI by regulating the IRE-1 signaling pathway via promoting the dissociation of GRP94 from IRE-1. This may provide a theoretical basis and a potential therapeutic target for clinical improvement of neural function recovery after SCI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human HAX1
Preise und Verfügbarkeit sind derzeit nicht verfügbar.