Merck
  • Chalcones and other compounds from the exudates of Angelica keiskei and their cancer chemopreventive effects.
Chalcones and other compounds from the exudates of Angelica keiskei and their cancer chemopreventive effects.
Journal of natural products (2006-01-31)
Toshihiro Akihisa, Harukuni Tokuda, Daisuke Hasegawa, Motohiko Ukiya, Yumiko Kimura, Fumio Enjo, Takashi Suzuki, Hoyoku Nishino
ZUSAMMENFASSUNG
Three new chalcones, xanthoangelol I (1), xanthoangelol J (2), and deoxydihydroxanthoangelol H (3), were isolated from an ethyl acetate-soluble fraction of exudates of the stems of Angelica keiskei, and their structures were established on the basis of spectroscopic methods. Nine aromatic compounds of known structure, 4-12, and a diacetylene, 13, were also isolated and identified from this same fraction. On evaluation of these compounds for their inhibitory effects on the induction of Epstein-Barr virus early antigen (EBV-EA) by 12-O-tetradecanoylphorbol-13-acetate (TPA) in Raji cells, 1, 2, 4, and 9-12 showed potent inhibitory effects on EBV-EA induction. In addition, upon evaluation of the inhibitory effects against activation of (+/-)-(E)-methyl-2[(E)-hydroxyimino]-5-nitro-6-methoxy-3-hexemide (NOR 1), a nitrogen oxide (NO) donor, six compounds, namely, 1, 2, 4, 9, 11, and 12, exhibited potent inhibitory effects. Further, isobavachalcone (4) exhibited inhibitory effects on skin tumor promotion in an in vivo two-stage mouse skin carcinogenesis test using 7,12-dimethylbenz[a]anthracene (DMBA) as an initiator and TPA as a promoter.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Xanthotoxin, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.