Merck
  • Identification and characterization of a new metallo-beta-lactamase, IND-5, from a clinical isolate of Chryseobacterium indologenes.
Identification and characterization of a new metallo-beta-lactamase, IND-5, from a clinical isolate of Chryseobacterium indologenes.
Antimicrobial agents and chemotherapy (2007-05-02)
Mariagrazia Perilli, Bibiana Caporale, Giuseppe Celenza, Cristina Pellegrini, Jean Denis Docquier, Marilina Mezzatesta, Gian Maria Rossolini, Stefania Stefani, Gianfranco Amicosante
ZUSAMMENFASSUNG
A new natural IND-type metallo-beta-lactamase variant, IND-5, was identified in a clinical isolate of Chryseobacterium indologenes. IND-5 shared 92.8% and 92.4% amino acid homology with IND-1 and IND-3, respectively. Purified enzyme (pI = 8.8, M(r) = 25,000) was able to hydrolyze penicillins, some narrow- and expanded-spectrum cephalosporins, and carbapenems but not monobactams.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
1,10-Phenanthrolin, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
2,6-Pyridindicarbonsäure, 99%
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Supelco
2,6-Pyridindicarbonsäure, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)