Merck
  • Three-dimensional quantitative structure-activity relationship analysis of human CYP51 inhibitors.
Three-dimensional quantitative structure-activity relationship analysis of human CYP51 inhibitors.
Drug metabolism and disposition: the biological fate of chemicals (2006-12-30)
Sean Ekins, Dayna C Mankowski, Dennis J Hoover, Michael P Lawton, Judith L Treadway, H James Harwood
ZUSAMMENFASSUNG
CYP51 fulfills an essential requirement for all cells, by catalyzing three sequential mono-oxidations within the cholesterol biosynthesis cascade. Inhibition of fungal CYP51 is used as a therapy for treating fungal infections, whereas inhibition of human CYP51 has been considered as a pharmacological approach to treat dyslipidemia and some forms of cancer. To predict the interaction of inhibitors with the active site of human CYP51, a three-dimensional quantitative structure-activity relationship model was constructed. This pharmacophore model of the common structural features of CYP51 inhibitors was built using the program Catalyst from multiple inhibitors (n = 26) of recombinant human CYP51-mediated lanosterol 14alpha-demethylation. The pharmacophore, which consisted of one hydrophobe, one hydrogen bond acceptor, and two ring aromatic features, demonstrated a high correlation between observed and predicted IC(50) values (r = 0.92). Validation of this pharmacophore was performed by predicting the IC(50) of a test set of commercially available (n = 19) and CP-320626-related (n = 48) CYP51 inhibitors. Using predictions below 10 microM as a cutoff indicative of active inhibitors, 16 of 19 commercially available inhibitors (84%) and 38 of 48 CP-320626-related inhibitors (79.2%) were predicted correctly. To better understand how inhibitors fit into the enzyme, potent CYP51 inhibitors were used to build a Cerius(2) receptor surface model representing the volume of the active site. This study has demonstrated the potential for ligand-based computational pharmacophore modeling of human CYP51 and enables a high-throughput screening system for drug discovery and data base mining.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Arachidonsäure, >95.0% (GC)
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Benzimidazol, 98%
Sigma-Aldrich
Arachidonsäure, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Clotrimazol
Sigma-Aldrich
Progesteron-Wasser, löslich, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Ketoconazol, 99.0-101.0% (EP, titration), meets EP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
2-Mercapto-1-methylimidazol, ≥99%
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Lanosterin, ≥93%, powder
Sigma-Aldrich
Sulfaphenazole, ≥98%
Supelco
Tolbutamid, analytical standard
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Supelco
Methimazol, analytical standard
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Testosteron -Lösung, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Supelco
Testosteron, VETRANAL®, analytical standard
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.