Merck
Topography, clinical, and genomic correlates of 5q myeloid malignancies revisited.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2012-03-01)
Andres Jerez, Lukasz P Gondek, Anna M Jankowska, Hideki Makishima, Bartlomiej Przychodzen, Ramon V Tiu, Christine L O'Keefe, Azim M Mohamedali, Denise Batista, Mikkael A Sekeres, Michael A McDevitt, Ghulam J Mufti, Jaroslaw P Maciejewski
ZUSAMMENFASSUNG
Interstitial deletions of chromosome 5q are common in acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS), pointing toward the pathogenic role of this region in disease phenotype and clonal evolution. The higher level of resolution of single-nucleotide polymorphism array (SNP-A) karyotyping may be used to find cryptic abnormalities and to precisely define the topographic features of the genomic lesions, allowing for more accurate clinical correlations. We analyzed high-density SNP-A karyotyping at diagnosis for a cohort of 1,155 clinically well-annotated patients with malignant myeloid disorders. results: We identified chromosome 5q deletions in 142 (12%) of 1,155 patients and uniparental disomy segments (UPD) in four (0.35%) of 1,155 patients. Patients with deletions involving the centromeric and telomeric extremes of 5q have a more aggressive disease phenotype and additional chromosomal lesions. Lesions not involving the centromeric or telomeric extremes of 5q are not exclusive to 5q- syndrome but can be associated with other less aggressive forms of MDS. In addition, larger 5q deletions are associated with either del(17p) or UPD17p. In 31 of 33 patients with del(5q) AML, either a deletion involving the centromeric and/or telomeric regions or heterozygous mutations in NPM1 or MAML1 located in 5q35 were present. Our results suggest that the extent of the affected region on 5q determines clinical characteristics that can be further modified by heterozygous mutations present in the telomeric extreme.