Merck
  • Does the FSHB c.-211G>T polymorphism impact Sertoli cell number and the spermatogenic potential in infertile patients?
Does the FSHB c.-211G>T polymorphism impact Sertoli cell number and the spermatogenic potential in infertile patients?
Andrology (2020-02-26)
Maria Schubert, Sophie Kaldewey, Lina Pérez Lanuza, Henrike Krenz, Martin Dugas, Sven Berres, Sabine Kliesch, Joachim Wistuba, Jörg Gromoll
ZUSAMMENFASSUNG
A genetic variant within the FSHB gene can deviate FSH action on spermatogenesis. The c.-211G>T FSHB single nucleotide polymorphism impacts FSHB transcription and biosynthesis due to interference with the LHX3 transcription factor binding. This SNP was previously shown to be strongly associated with lowered testicular volume, reduced sperm counts, and decreased FSH levels in patients carrying one or two T-alleles. To determine the impact of the SNP FSHB c.-211G>T on Sertoli cell (SC) number, Sertoli cell workload (SCWL) and thereby spermatogenic potential. Testicular biopsies of 31 azoospermic, homozygous T patients (26 non-obstructive azoospermia (NOA), and five obstructive azoospermia (OA)) were matched to patients with GG genotype. Marker proteins for SC (SOX9), spermatogonia (MAGE A4), and round spermatids (CREM) were used for semi-automatical quantification by immunofluorescence. SCWL (number of germ cells served by one SC) was determined and an unbiased clustering on the patient groups performed. Quantification of SC number in NOA patients did not yield significant differences when stratified by FSHB genotype. SC numbers are also not significantly different between FSHB genotypes for the OA patient group and between NOA and OA groups. SCWL in the NOA patient cohort is significantly reduced when compared to the OA control patients; however, in neither group an effect of the genotype could be observed. The cluster analysis of the whole study cohort yielded two groups only, namely NOA and OA, and no clustering according to the FSHB genotype. The FSHB c.-211G>T polymorphism does not affect SC numbers or SCWL, thereby in principle maintaining the spermatogenic potential. The previously observed clinical phenotype for the FSHB genotype might therefore be caused by a hypo-stimulated spermatogenesis and not due to a decreased SC number.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-CREM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.