Merck
Synthesis and biological activities of some new fluorinated coumarins and 1-aza coumarins.
European journal of medicinal chemistry (2007-10-26)
Rajesh G Kalkhambkar, Geeta M Kulkarni, Chandrappa M Kamanavalli, N Premkumar, S M B Asdaq, Chung Ming Sun
ZUSAMMENFASSUNG
A series of new fluorinated coumarins and 1-aza coumarins have been synthesized and the presence of fluorine in these molecules and its effect on their anti-microbial, anti-inflammatory and analgesic activities are discussed. The results of bioassay showed that these newly synthesized compounds containing fluorine exhibit moderate analgesic and excellent anti-inflammatory and potential anti-bacterial and anti-fungal activities, compared to the other halogenated compounds. All the newly synthesized compounds were characterized by elemental analysis, IR, 1H NMR, 13C NMR, 19F NMR, EI-MS, and FAB-MS. The ORTEP diagram of one of the compounds is reported herein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard