Merck
Serine ADP-Ribosylation Depends on HPF1.
Molecular cell (2017-02-14)
Juan José Bonfiglio, Pietro Fontana, Qi Zhang, Thomas Colby, Ian Gibbs-Seymour, Ilian Atanassov, Edward Bartlett, Roko Zaja, Ivan Ahel, Ivan Matic
ZUSAMMENFASSUNG
ADP-ribosylation (ADPr) regulates important patho-physiological processes through its attachment to different amino acids in proteins. Recently, by precision mapping on all possible amino acid residues, we identified histone serine ADPr marks in the DNA damage response. However, the biochemical basis underlying this serine modification remained unknown. Here we report that serine ADPr is strictly dependent on histone PARylation factor 1 (HPF1), a recently identified regulator of PARP-1. Quantitative proteomics revealed that serine ADPr does not occur in cells lacking HPF1. Moreover, adding HPF1 to in vitro PARP-1/PARP-2 reactions is necessary and sufficient for serine-specific ADPr of histones and PARP-1 itself. Three endogenous serine ADPr sites are located on the PARP-1 automodification domain. Further identification of serine ADPr on HMG proteins and hundreds of other targets indicates that serine ADPr is a widespread modification. We propose that O-linked protein ADPr is the key signal in PARP-1/PARP-2-dependent processes that govern genome stability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-GAPDH-Maus-mAb (6C5), liquid, clone 6C5, Calbiochem®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
PARP-Inhibitor, 3-ABA, The PARP Inhibitor I, 3-ABA, also referenced under CAS 3544-24-9, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.