Merck
  • Side chain flexibilities in the human ether-a-go-go related gene potassium channel (hERG) together with matched-pair binding studies suggest a new binding mode for channel blockers.
Side chain flexibilities in the human ether-a-go-go related gene potassium channel (hERG) together with matched-pair binding studies suggest a new binding mode for channel blockers.
Journal of medicinal chemistry (2009-06-19)
Ulrich Zachariae, Fabrizio Giordanetto, Andrew G Leach
ZUSAMMENFASSUNG
The cardiac hERG K(+) channel constitutes a long-standing and expensive antitarget for the drug industry. From a study of the flexibility of hERG around its internal binding cavity, we have developed a new structural model of drug binding to hERG, which involves binding orthogonal to the pore channel and therefore can exploit the up to 4-fold symmetry of the tetrameric channel. This binding site has a base formed by four tyrosine side chains that complement reported ligand-based pharmacophores. The model is able to rationalize reduced hERG potency in matched molecular pair studies and suggests design guidelines to optimize against hERG not relying simply on lipophilicity reduction. The binding model also suggests a molecular mechanism for the link between high-affinity hERG binding and C-type inactivation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Clozapin-N-Oxid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ketoconazol, 99.0-101.0% (EP, titration), meets EP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
(−)-Nikotin, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Terfenadin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
(−)-Nikotin -Lösung, 1.0 mg/mL, analytical standard, for drug analysis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N-Desmethylclozapine
Preise und Verfügbarkeit sind derzeit nicht verfügbar.