Merck
  • COVID-19 severity correlates with airway epithelium-immune cell interactions identified by single-cell analysis.
COVID-19 severity correlates with airway epithelium-immune cell interactions identified by single-cell analysis.
Nature biotechnology (2020-06-28)
Robert Lorenz Chua, Soeren Lukassen, Saskia Trump, Bianca P Hennig, Daniel Wendisch, Fabian Pott, Olivia Debnath, Loreen Thürmann, Florian Kurth, Maria Theresa Völker, Julia Kazmierski, Bernd Timmermann, Sven Twardziok, Stefan Schneider, Felix Machleidt, Holger Müller-Redetzky, Melanie Maier, Alexander Krannich, Sein Schmidt, Felix Balzer, Johannes Liebig, Jennifer Loske, Norbert Suttorp, Jürgen Eils, Naveed Ishaque, Uwe Gerd Liebert, Christof von Kalle, Andreas Hocke, Martin Witzenrath, Christine Goffinet, Christian Drosten, Sven Laudi, Irina Lehmann, Christian Conrad, Leif-Erik Sander, Roland Eils
ZUSAMMENFASSUNG
To investigate the immune response and mechanisms associated with severe coronavirus disease 2019 (COVID-19), we performed single-cell RNA sequencing on nasopharyngeal and bronchial samples from 19 clinically well-characterized patients with moderate or critical disease and from five healthy controls. We identified airway epithelial cell types and states vulnerable to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection. In patients with COVID-19, epithelial cells showed an average three-fold increase in expression of the SARS-CoV-2 entry receptor ACE2, which correlated with interferon signals by immune cells. Compared to moderate cases, critical cases exhibited stronger interactions between epithelial and immune cells, as indicated by ligand-receptor expression profiles, and activated immune cells, including inflammatory macrophages expressing CCL2, CCL3, CCL20, CXCL1, CXCL3, CXCL10, IL8, IL1B and TNF. The transcriptional differences in critical cases compared to moderate cases likely contribute to clinical observations of heightened inflammatory tissue damage, lung injury and respiratory failure. Our data suggest that pharmacologic inhibition of the CCR1 and/or CCR5 pathways might suppress immune hyperactivation in critical COVID-19.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulbeccos Phosphatgepufferte Kochsalzlösung, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Roche
Lysepuffer für Erythrozyten, solution, Roche, pkg of 100 mL, sufficient for 50-500 reactions