Merck
  • Extensive Plasmid Library to Prepare Tau Protein Variants and Study Their Functional Biochemistry.
Extensive Plasmid Library to Prepare Tau Protein Variants and Study Their Functional Biochemistry.
ACS chemical neuroscience (2020-08-25)
Thomas K Karikari, Sophie Keeling, Emily Hill, Juan Lantero Rodrı Guez, David A Nagel, Bruno Becker, Kina Höglund, Henrik Zetterberg, Kaj Blennow, Eric J Hill, Kevin G Moffat
ZUSAMMENFASSUNG
Tau neurofibrillary tangles are key pathological features of Alzheimer's disease and other tauopathies. Recombinant protein technology is vital for studying the structure and function of tau in physiology and aggregation in pathophysiology. However, open-source and well-characterized plasmids for efficiently expressing and purifying different tau variants are lacking. We generated 44 sequence-verified plasmids including those encoding full length (FL) tau-441, its four-repeat microtubule-binding (K18) fragment, and their respective selected familial pathological variants (N279K, V337M, P301L, C291R, and S356T). Moreover, plasmids for expressing single (C291A), double (C291A/C322A), and triple (C291A/C322A/I260C) cysteine-modified variants were generated to study alterations in cysteine content and locations. Furthermore, protocols for producing representative tau forms were developed. We produced and characterized the aggregation behavior of the triple cysteine-modified tau-K18, often used in real-time cell internalization and aggregation studies because it can be fluorescently labeled on a cysteine outside the microtubule-binding core. Similar to the wild type (WT), triple cysteine-modified tau-K18 aggregated by progressive β-sheet enrichment, albeit at a slower rate. On prolonged incubation, cysteine-modified K18 formed paired helical filaments similar to those in Alzheimer's disease, sharing morphological phenotypes with WT tau-K18 filaments. Nonetheless, cysteine-modified tau-K18 filaments were significantly shorter (p = 0.002) and mostly wider than WT filaments, explainable by their different principal filament elongation pathways: vertical (end-to-end) and lateral growth for WT and cysteine-modified, respectively. Cysteine rearrangement may therefore induce filament polymorphism. Together, the plasmid library, the protein production methods, and the new insights into cysteine-dependent aggregation should facilitate further studies and the design of antiaggregation agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Roche
DNase I, rekombinant, RNase-frei, from bovine pancreas, expressed in Pichia pastoris
Sigma-Aldrich
Heparin Natriumsalz aus Schweinedarm, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane