Merck
  • Case of fatal Blastoschizomyces capitatus infection occurring in a patient receiving empiric micafungin therapy.
Case of fatal Blastoschizomyces capitatus infection occurring in a patient receiving empiric micafungin therapy.
Antimicrobial agents and chemotherapy (2009-09-10)
Paul Chittick, Elizabeth L Palavecino, Brian Delashmitt, James Evans, James E Peacock
ZUSAMMENFASSUNG
We report the first case, to our knowledge, of Blastoschizomyces capitatus infection occurring in a patient receiving empirical echinocandin therapy for neutropenic fevers. Clinicians should consider B. capitatus infection in those neutropenic patients who remain febrile despite echinocandin therapy or who develop yeast bloodstream infections while receiving an echinocandin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorcytosin, nucleoside analog
Preise und Verfügbarkeit sind derzeit nicht verfügbar.