Merck
  • Peripheral Markers of Neurovascular Unit Integrity and Amyloid-β in the Brains of Menopausal Women.
Peripheral Markers of Neurovascular Unit Integrity and Amyloid-β in the Brains of Menopausal Women.
Journal of Alzheimer's disease : JAD (2021-02-09)
Muthuvel Jayachandran, Virginia M Miller, Brian D Lahr, Kent R Bailey, Val J Lowe, Julie A Fields, Michelle M Mielke, Kejal Kantarci
ZUSAMMENFASSUNG
The identification of blood-borne biomarkers for the diagnosis and prognosis of Alzheimer's disease and related dementias is more feasible at the population level than obtaining cerebrospinal fluid or neuroimaging markers. This study determined the association of blood microvesicles, derived from cells of the neurovascular unit, with brain amyloid-β deposition in menopausal women. A subset of women from the Kronos Early Estrogen Prevention Study underwent brain amyloid-β positron emission tomography three years following cessation of study treatment with placebo (PL, n = 29), transdermal 17β-estradiol (tE2; n = 21), or oral conjugated equine estrogen (oCEE; n = 17). Isolated peripheral venous blood microvesicles were analyzed by digital flow cytometry using fluorophore conjugated antibodies directed toward total tau, amyloid-β 1-42 (Aβ1-42), neuron specific class III β-tubulin (Tuj1), microglia ionized calcium -binding adaptor molecule 1(Iba1), glial fibrillary acid protein (GFAP), and low density lipoprotein receptor-related protein1 (LRP1). Principal components analysis reduced the dimensionality of these selected six markers to two principal components (PCs). Proportional odds ordinal logistic regression analysis was used with amyloid-β deposition regressed on these PCs. Only the number of microvesicles positive for Aβ1-42 differed statistically among prior treatment groups (median [IQR]: 6.06 [2.11, 12.55] in PL; 2.49 [0.73, 3.59] in tE2; and 4.96 [0.83, 10.31] in oCEE; p = 0.032). The joint association between the 2 PCs and brain amyloid-β deposition was significant (p = 0.045). Six selected markers expressing peripheral blood microvesicles derived from cells of the neurovascular unit, when summarized into two principal components, were associated with brain amyloid-β deposition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PiB, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.