Merck
  • Multimerized self-assembled caged two-in-one siRNA nanoparticles for photomodulation of RNAi-induced gene silencing.
Multimerized self-assembled caged two-in-one siRNA nanoparticles for photomodulation of RNAi-induced gene silencing.
Chemical science (2021-06-08)
Changmai Chen, Nannan Jing, Zhongyu Wang, Yu Zhang, Wei Chen, Xinjing Tang
ZUSAMMENFASSUNG
We rationally designed and developed caged siRNA nanoparticles (Multi-Chol-siRNA) self-assembled with cholesterol-modified multimerized caged siRNAs for photomodulation of siRNA gene silencing activity. Strong resistance to serum nuclease and RNase A was observed for these cholesterol-modified caged siRNA nanoparticles due to the formation of nanostructures with high intensity of siRNA. These caged Multi-Chol-siRNA self-assembled nanoparticles were successfully used to achieve photochemical regulation of both exogenous GFP and endogenous Eg5 gene expressions with a GFP/RFP transient transfection system and Eg5-associated assays, respectively. Further, Two-in-One caged Multi-Chol-siGFP/siEg5 self-assembled nanoparticles simultaneously targeting GFP and Eg5 genes were also developed. The caged Multi-Chol-siRNA self-assembled nanoparticles have demonstrated the effectiveness of enhancing photomodulation of multiple RNAi-induced gene silencing activities in cells.